2-(4-Chlorophenoxy)nicotinic acid ≥98%

Supplier: Thermo Scientific
Warning

A14359.03 A14359.06
ALFAA14359.03EA 0 SGD
ALFAA14359.03 ALFAA14359.06
2-(4-Chlorophenoxy)nicotinic acid ≥98%
2-(4-Chlorophenoxy)nicotinic acid

Purity: 98% CAS No.: 51362-37-9 Molecular Formula: C12H8ClNO3 Formula Weight: 249.64 Melting Point: 157-159° MDL No.: MFCD00052116

Formula: C₁₂H₈ClNO₃
MW: 249,64 g/mol
Melting Pt: 157…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00052116
CAS Number: 51362-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR