Thionicotinamide ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Pyridine-3-carbothioamide

A11144.14 A11144.22
ALFAA11144.14EA 0 SGD
ALFAA11144.14 ALFAA11144.22
Thionicotinamide ≥98%
Thionicotinamide

Purity: 98% CAS No.: 4621-66-3 Molecular Formula: C6H6N2S Formula Weight: 138.19 Flash Point: 160°(320°F) Melting Point: ca 190° dec. Beilstein Reference: 109593 MDL No.: MFCD00006399

Formula: C₆H₆N₂S
MW: 138,19 g/mol
Melting Pt: ∼ 190 °C (dec.)
Flash Pt: 160 °C (320 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006399
CAS Number: 4621-66-3
EINECS: 225-036-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR