2-Methyl-4-pyridineboronic acid pinacol ester, (max. 3% H₂O) 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Methylpyridine-4-boronic acid Pinacol ester, 2-(2-Methyl-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H27450.MD H27450.03
ALFAH27450.MDEA 0 SGD
ALFAH27450.MD ALFAH27450.03
2-Methyl-4-pyridineboronic acid pinacol ester, (max. 3% H₂O) 95%
2-Methyl-4-pyridineboronic acid pinacol ester

Purity: 95% CAS No.: 660867-80-1 Molecular Formula: C12H18BNO2 Formula Weight: 219.09 Storage Temperature: -30°C to -10°C Melting Point: 66-71° MDL No.: MFCD08061590

Formula: C₁₂H₁₈BNO₂
MW: 219.09 g/mol
Melting Pt: 66…71 °C
Storage Temperature: Freezer
MDL Number: MFCD08061590
CAS Number: 660867-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR