2-Benzyloxy-5-pyridineboronic acid 95%

Supplier: Thermo Scientific
Warning

Synonyms: 6-Benzyloxypyridine-3-boronic acid, 6-(Benzyloxy)pyridine-3-boronic Acid

H27346.MD H27346.03
ALFAH27346.MDEA 0 SGD
ALFAH27346.MD ALFAH27346.03
2-Benzyloxy-5-pyridineboronic acid 95%
2-Benzyloxy-5-pyridineboronic acid

Purity: 95% CAS No.: 929250-35-1 Molecular Formula: C12H12BNO3 Formula Weight: 229.04 Melting Point: 107-110° MDL No.: MFCD06798262

Formula: C₁₂H₁₂BNO₃
MW: 229,04 g/mol
Melting Pt: 107…110 °C
Storage Temperature: Ambient
MDL Number: MFCD06798262
CAS Number: 929250-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR