2-Aminoisonicotinic acid 96%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Amino-4-pyridinecarboxylic acid, 2-Aminopyridine-4-carboxylic acid

L19845.06 L19845.14
ALFAL19845.06EA 0 SGD
ALFAL19845.06 ALFAL19845.14
2-Aminoisonicotinic acid 96%
2-Aminoisonicotinic acid

Purity: 96% CAS No.: 13362-28-2 Molecular Formula: C6H6N2O2 Formula Weight: 138.13 Odour: Odourless Melting Point: 300-302° Beilstein Reference: 3156 MDL No.: MFCD00233714

Formula: C₆H₆N₂O₂
MW: 138,13 g/mol
Melting Pt: 300…302 °C
Storage Temperature: Ambient
MDL Number: MFCD00233714
CAS Number: 13362-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR