Cobalt(II) oxalate, anhydrous 98%
Supplier: Thermo Scientific
|
Danger
|
B22956.09
B22956.18
ALFAB22956.09EA
110
SGD
ALFAB22956.09
ALFAB22956.18
Cobalt(II) oxalate, anhydrous 98%
Cobalt (II) oxalate
Purity: 98%
CAS No.: 814-89-1
Molecular Formula: C2CoO4
Formula Weight: 146.95
Melting Point:
Formula:
C₂CoO₄ MW: 146,95 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00050793 CAS Number: 814-89-1 EINECS: 212-409-3 Merck Index: 14,2445 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...