3-(Benzyloxy)-4-methoxybenzaldehyde ≥98%

Supplier: Thermo Scientific

Synonyms: 3-Benzyloxy-4-methoxybenzaldehyde

L06830.09 L06830.18
ALFAL06830.09EA 0 SGD
ALFAL06830.09 ALFAL06830.18
3-(Benzyloxy)-4-methoxybenzaldehyde ≥98%
3-(Benzyloxy)-4-methoxybenzaldehyde

Purity: 98% CAS No.: 6346-05-0 Molecular Formula: C15H14O3 Formula Weight: 242.27 Melting Point: 60-64° Beilstein Reference: 794627

Formula: C₁₅H₁₄O₃
MW: 242,27 g/mol
Melting Pt: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00003386
CAS Number: 6346-05-0
EINECS: 228-752-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR