VWR® TraceClean® Straight Side Glass Jars, Clear
Supplier: VWR Collection
89126-578 89126-596
VWRU89126-578EA 800 SGD
VWRU89126-578 VWRU89126-596
VWR® TraceClean® Straight Side Glass Jars, Clear
Jars General Use Jars
These TraceClean® glass jars are used primarily for the collection of soil, sediments, and slurry media for testing Semi-volatiles, Pesticides, PCBs, Diesel Range Organics, TPH, Metals, and Phenolics.
  • Clear glass fabrication
  • Straight-sided design
  • Used as a containment vessel
  • Suited to the collection of inorganic samples
Secondary uses include containment, bench vessels, and archiving.
The 950 ml clear jar 89094-235 contains 5 ml (1:1) of the chemical preservative H₂SO₄.
Quality Assured® with VWR® Logo Sample ID Label products, a lot certificate
More Product Information