Whatman™ Filters for Water, Air and Soil Testing
Accurate and reliable analytical results are important when monitoring air, water and soil for Environmental testing. GE knows that you need to be sure your filtration step is efficient, reproducible and preserves the integrity of your sample. Whatever you are using one of our glass papers, syringe filters or other Whatman™ products, you can count on GE to understand that quality matters.
Whatman™ Filters for Water Testing
With Whatman™ filters you can test your samples according to the environmental water testing requirements. The following testing have to be performed:
|
Whatman™ has developed specific filters to guarantee reliable results and to let you perform easily each analysis even starting from critical samples.
For example, with the 934-AH RTU glass fiber filters, which are supplied in a prewashed and preweighed format, enable a considerable time saving when performing suspended solid analysis.
The GD/XP syringe filters, including an integrated prefiltration with dual-layer prefilter stack and one final 0.45 micron membrane, allow an easy filtration of hard-to-filter samples. Futhermore the pre-filter in polypropylene minimizes ion extractables making them suitable for metal trace analysis by ICP-MS. Find out how Anotop™ IC, Puradisc Aqua 30, Puradisc, Spartan, GD/XP and GD/X™ and Mini-Uniprep can help you!
Customer testimonial: Whatman™ filters for Water Quality Testing