VWR Chemicals
VWR Chemicals are our own wide range of laboratory and industrial chemicals with guaranteed high quality, detailed specifications, safe packaging and excellent service offering great value for money.
Everything from basic technical reagents to high purity chemicals for trace analysis plus extensive speciality ranges for applications like microbiology, cell diagnostics and life sciences.
From HPLC, GC and IC through TLC, preparative and flash chromatography, VWR offers the most extensive ranges of columns, reagents and solvents for chromatographers in all industries. The VWR Collection also offers an extensive range of vials, syringes and other consumables to make separations easier!
From the most exacting sample preparation with NORMATOM® high purity acids, specialist ranges tailored to specific analysis including NMR. Karl Fischer and AAS to ARISTAR® ICP/ICP-MS and AVS® TITRINORM® IC standards.
VWR Chemicals offers over 400 reagents suitable for many inorganic and analytical applications. The specifications quote guaranteed maximum permitted limits of impurity and have a 98% stock availability. Choose from AnalaR® NORMAPUR® for analytical work, RECTAPUR ® for everyday tasks and TECHNICAL® for the basics to the ultra high purity Ultrapure NORMATOM® for almost invisible sample preparation in trace analysis. Speciality products such as Kjeldahl catalysts, Karl Fischer titrants as well as products compliant with ACS, Reag.Ph. Eur. and USP requirements are available.
- AnalaR® NORMAPUR® Analytical grade acids
- GPR RECTAPUR® General use acids
- TECHNICAL® Basic specification acids
- AnalaR® NORMAPUR® Analytical grade alkalis
- GPR RECTAPUR® General use alkalis
- TECHNICAL® Basic specification alkalis
- AnalaR® NORMAPUR® Analytical grade salts
- GPR RECTAPUR® General use salts
- TECHNICAL® Basic specification salts
- Kjeldahl catalysts
- Reag PH EUR Standard Solutions
- Indicators
Some key elements of laboratory management are safety, cleaning and lab auxiliairies. VWR offers an extensive choice of products for managing spills including the versatile Algosol as well as the basic spillage absorption granules. For housekeeping - Labwash offers a range of products for manual or machine washing . Some other interesting products will be visible as greases, drying agents.....
VWR offers a comprehensive range of cell diagnostic reagents including fixation, dehydration and embedding in paraffin wax. A full selection of stains and dyes plus mountants and immersion oils enable full visualisation of samples.