Accelerating Genomic Discoveries Through Innovation: New repliQa HiFi ToughMix
Webinar
Wednesday, 24th Febuary, 2021
10:00 to 11:00 SGT
Replay
In this webinar, Quantabio introduces the newest High Fidelity PCR master mix capable of fast PCR amplification while overcoming common PCR inhibitors. Review how scientists are applying repliQa HiFi ToughMix for the following applications in cloning, Protein Expression and RNA Therapeutics.
We will discuss: 
  • How scientists used repliQa HiFi Toughmix to increase yield while decreasing non-specificity from hard-to-digest plasmids. 
  • How scientists decreased amplification time using repliQa HiFi Toughmix for RNA therapeutics.
Featured Product 
The repliQa HiFi ToughMix x is a 2x, ready-to-use solution that contains all the components for high fidelity PCR amplification, including a genetically modified DNA polymerase coupled with hot start antibodies. 
Features and Benefits: 
  • High Fidelity - >90x wild type Taq
  • Fast Speed - 2–3x faster PCR results with extension rates as fast as 1 kb/sec*
  • Tough Tested − tolerant to a wide range of PCR inhibitors common in PCR crude samples
  • Superior yield and sensitivity
  • Long Amplification - +24 kb gDNA and +40 kb λ DNA
Potential Applications:
  • Cloning
  • RNA Therapeutics
  • Gene Expression Analysis
  • Genotyping
  • Mutagenesis
  • Long range PCR
  • Difficult template PCR