Sterile Solutions for Surfaces in Cleanrooms
The solutions are filtered to 0.2 µm before filling and packaging in cleanroom conditions, and are either aseptically filled or terminally irradiated.  
Neutral detergent
  • High-foaming liquid detergent, neutral pH
  • 0.2 µm filtered, gamma irradiated, double bagged
  • Supplied with certificates of analysis, irradiation and sterility
  • Supplied as: 100 x 30 ml or 50 x 50 ml concentrate
Ethanol and IPA
  • Broad spectrum disinfectant
  • Passes CEN 1276, CEN 1650 and CEN 13697
  • 0.2 µm filtered, gamma irradiated (900 ml) or aseptically filled (5 litres), double bagged
  • Supplied with certificates of analysis, sterility and irradiation, where applicable
  • Denatured Ethanol: 70% ethanol, 30% water (WFI) denatured with IPA
  • Isopropyl alcohol: 70% isopropyl alcohol, 30% water (WFI)
  • Also available as an aerosol with purified water
  • Available as: 6 x 900 ml spray bottle; 6 x 1 l spray bottle; 4 x 5 l or 2 x 5 l canister; 12 x 300 ml aerosol
Sporicidal disinfectants
  • Two-part chlorine dioxide generating system
  • Passes CEN 1276, CEN 1650, CEN 13697 and CEN 13704
  • 0.2 µm filtered, aseptically filled and double bagged
  • Supplied with certificates of analysis and sterility
  • Based on chlorine dioxide
  • Compatible in rotation with amphoteric disinfectants
  • Effective against spores within only 5-8 minutes!
  • Available as: 25 x (2 x 100 ml) concentrate;    
    4 x (4.9 l + 100 ml) ready-to-use
Amphoteric disinfectants
  • Highly effective surface active disinfectant
  • Passes CEN 1276, CEN 1650 and CEN 13697
  • 0.2 µm filtered, aseptically filled and double bagged
  • Supplied with certificates of analysis and sterility
  • Validated HPLC and Titration test methods available
  • Compatible in rotation with a sporicidal or QAC disinfectant
  • Available as: 50 x 50 ml or 50 x 100 ml concentrate;
    6 x 900 ml spray bottle; 4 x 5 l or 2 x 5 l canister  
Quaternary Ammonium disinfectants
  • Highly effective surface active disinfectant based on Quats
  • 0.2 µm filtered, aseptically filled and double bagged
  • Supplied with certificates of analysis and sterility
  • Highly compatible with materials
  • For a broad range of applications
  • Unit: 50 x 25 ml or 50 x 100 ml concentrate;
    6 x 900 ml spray bottle; 4 x 5 l or 2 x 5 l canister
Contact us