Quantus Overalls
Features:
  • Lapped seams for a well-sealed and secure seam covering
  • Plastic front zipper (double covered) 
  • High collar with press stud
  • Tunnel-guided rubber waistband; adjustable via button 
  • Comfortable raglan style
  • Antistatic knitted cuffs as sleeve closures
  • Width adjustable leg closures with press studs
  • Press studs on left breast; white PES clipband (W x H) 12 cm x 2 cm
  • Stitched collar patch with label
  • Hanger in collar
Optional Features:
  • Antistatic knitted cuffs at ankles
  • Clipband of cleanroom fabric (printed/unprinted)
  • Individual printing: Corporate logo, name, codes, etc.
  • Loop for attaching pens, mobile phones, IDs, etc. (Size: (W x H) 8 cm x 4 cm)
  • ESD functional models are available upon request
  • Combined Overall: Coverall with integrated flat hood including mesh ear inserts, open field of view, 4 press studs at the front for adjustment, hood can be adjusted at the back means of press studs
Sizes: XXS - XXXL, Special sizes available!