Beaker and flask holders, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F183090250
BELAF183090250EA 405 SGD
BELAF183090250
Beaker and flask holders, SP Bel-Art
Supports Flask Supports
Lead ring, with red vinyl coating and four stainless steel brackets. Holds beakers and flasks firmly in a fixed position. Ideal for use in water baths as it allows liquid to circulate beneath the vessel base.
  • Good chemical resistance
  • Good thermal resistance
  • Easy, simple handling
Order Now
SPECIFICATIONS
Environmentally Preferable