Water treatment system, Pure Aqua Service
Supplier: Binder
8012-0759
BIND8012-0759EA 2250 SGD
BIND8012-0759
Water treatment system, Pure Aqua Service
Water Purification Systems Deionisers
This system is used to fully desalinate faucet water for BINDER climate chamber humidification systems. The ion exchanger system consists of a pressure container, a single-use filter cartridge and a pressure container lid.
  • Can be used irrespective of drinking water quality
  • Robust system with safety pressure container
  • Reliable replacement of single-use cartridge when resin is used up
  • Operating condition of water treatment system is permanently displayed
The continuous water quality monitoring system is integrated in the lid of the Pure Aqua Service andMore Product Information