Drying ovens, FED 400
Supplier: Binder
9010-0216
BIND9010-0216EA 0 SGD
BIND9010-0216
Drying ovens, FED 400
Ovens Standard Ovens
Multi-functional drying ovens with stainless steel chambers which are ideal for use in the pharmaceutical and chemical industries, biotechnology, medicine and universities. Ovens feature one temperature program ramp, an adjustable temperature safety device, Class 2 (DIN 12880) with visual alarm and adjustable ventilation by means of front ventilation slide and rear 50 mm Ø exhaust duct. Fast recovery times after opening the door or air change. GLP/GMP compliance is assisted by the RS422 interface and by optional APT-COM™ DataControlSystem software.
  • APT.line™ preheating chamber
More Product Information