Evaporating dishes, PFA, BOLA
Supplier: Bohlender
A171-01
BOHLA171-01EA 281 SGD
BOHLA171-01
Evaporating dishes, PFA, BOLA
Trays/Dishes/Pots Evaporating Basins/Dishes
Evaporating dishes, made of PFA, translucent, with spout.
  • Universal chemical tesistance
  • Temperature resistance: from −200 to +250 °C
  • Non-porous