Thermometer for distillation processes, BOLA
Supplier: Bohlender
B290-03
BOHLB290-03EA 634 SGD
BOHLB290-03
Thermometer for distillation processes, BOLA
Thermometers Analogue Thermometers
Distillation thermometer made of PTFE jacketed glass. Suitable for 500 to 6000 ml reaction vessels.