Condensers, Liebig, vacuum, BOLA
Supplier: Bohlender
B295-02 B295-04
BOHLB295-02EA 1540 SGD
BOHLB295-02 BOHLB295-04
Condensers, Liebig, vacuum, BOLA
Condensers
PFA, PTFE, PP and glass.
  • Temperature resistance from −200 to +250 °C
  • Very good chemical resistance
  • Transparency: transparent
One-piece cooling tube with ground joint socket and cone made of PTFE, cooling jacket made of borosilicate glass with hose connectors made of PP and nuts for connection of cooling water. The distillate is only exposed to PFA/PTFE. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.