Magnetic stirring bar set, with retriever, BOLA
Supplier: Bohlender
C348-10
BOHLC348-10EA 222 SGD
BOHLC348-10
Magnetic stirring bar set, with retriever, BOLA
Stirrers Magnetic Stirring Bars
Magnetic stirring bar set containing the most common magnetic stirring bars and a retriever.
  • Chemical resistant
  • Ideal for beginners, for testing different kinds and dimensions of magnetic stirring bars
Temperature resistance: −200 to +250 °C.
L×W×H of box: 175×110×30 mm
Delivery information: Set consists of one each of (length×Ø): Cylindrical 10×6, 15×4,5, 20×6, 25×6, 30×6, 40×8, 50×8, 60×9 mm; Pivot ring 15×5, 25×6, 40×8 mm; Triangular 25×8, 40×14 mm; and 150 mm retriever.