Stirrer shafts, double-moon shape, BOLA
Supplier: Bohlender
C374-12 C374-18 C374-14
BOHLC374-12EA 721 SGD
BOHLC374-12 BOHLC374-18 BOHLC374-14
Stirrer shafts, double-moon shape, BOLA
Stirrers Stirring Shafts
Stirrer shafts made of PTFE-jacketed stainless steel, two each tilting half-moon stirrer blades with double-sided groove and access for the stirrer shaft completely made of PTFE.
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C
  • Creates tangential flow with little turbulence
  • With chucking diameter of 8 mm
Half-moon blade is ideal for stirring in round-bottom flasks with ground joint neck.
Order Now
SPECIFICATIONS
Environmentally Preferable