Stirring shafts, two paddles, BOLA
Supplier: Bohlender
C380-02 C380-10 C380-16 C380-08 C380-04 C380-18 C380-14 C380-12
BOHLC380-02EA 1220 SGD
BOHLC380-02 BOHLC380-10 BOHLC380-16 BOHLC380-08 BOHLC380-04 BOHLC380-18 BOHLC380-14 BOHLC380-12
Stirring shafts, two paddles, BOLA
Stirrers Stirring Shafts
Stirrer shafts made of PTFE-jacketed stainless steel and the paddles made of PTFE.
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C
Two paddles arranged crosswise at 90°. Upper paddle is fixed by means of clamp screws made of PEEK compound.
Order Now
SPECIFICATIONS
Environmentally Preferable