Stirrer blades, moon-shaped, with angular groove
Supplier: Bohlender
C401-02 C401-04 C401-08
BOHLC401-02EA 20.3 SGD
BOHLC401-02 BOHLC401-04 BOHLC401-08
Stirrer blades, moon-shaped, with angular groove
Stirrers Stirring Blades
Stirrer blades made of PTFE, with angular groove and 5,8 mm bore diameter.
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C
Suitable for vessels with a 29/32 ground joint and glass stirrer shafts and KPG glass stirrer shafts.