Stirrer shafts, with four bladed propeller, BOLA
Supplier: Bohlender
C484-18 C484-36 C484-32 C484-34 C484-22 C484-40 C484-44 C484-50
BOHLC484-18EA 841 SGD
BOHLC484-18 BOHLC484-36 BOHLC484-32 BOHLC484-34 BOHLC484-22 BOHLC484-40 BOHLC484-44 BOHLC484-50
Stirrer shafts, with four bladed propeller, BOLA
Stirrers Stirring Shafts
Stirrer shafts made of PTFE-jacketed stainless steel and the propeller completely made of PTFE.
  • With 45° angled angular blades
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C
Order Now
SPECIFICATIONS
Environmentally Preferable