Stirrer shafts for magnetic stirrer heads, glass, BOLA
Supplier: Bohlender
C492-16 C492-24
BOHLC492-16EA 357 SGD
BOHLC492-16 BOHLC492-24
Stirrer shafts for magnetic stirrer heads, glass, BOLA
Stirrers Stirring Shafts
Stirrer shafts made of borosilicate glass with integrated counterbores for the three stud screws of the rotor of the magnetic stirrer head.
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C
Ideal for use together with stirrer blades which can be fixed individually.
Order Now
SPECIFICATIONS
Environmentally Preferable