Multiple distributors for bottles with GL 45 thread, BOLA
Supplier: Bohlender
D618-48
BOHLD618-48EA 593 SGD
BOHLD618-48
Multiple distributors for bottles with GL 45 thread, BOLA
Closures Bottle Closures
PPS screw cap, black, distributor body made from PTFE.
  • Excellent chemical resistance
  • Temperature resistant from –200 to +250 °C
  • Suitable for vacuum applications
For the removal or filling of aggressive or pure liquids. For the connection of hoses, tubes and probes into vessels. Hoses with different external diameters can be inserted through the necks.
Order Now
SPECIFICATIONS
Environmentally Preferable