Screw caps for sensors, BOLA
Supplier: Bohlender
D780-14
BOHLD780-14EA 209 SGD
BOHLD780-14
Screw caps for sensors, BOLA
Closures Bottle Closures
PPS/PTFE.
  • Temperature resistance: from 0 to +110 °C
  • Universal chemical resistance
  • For contamination-free insertion of sensors into bottles with GL 45 thread
Black screw cap made of PPS with GL 45 thread, movable body made of PTFE with adjusting screw for fixing and sealing sensors. The product is only exposed to PTFE.
Order Now
SPECIFICATIONS
Environmentally Preferable