Flexible distributor for bottles, BOLA
Supplier: Bohlender
D804-12
BOHLD804-12EA 275 SGD
BOHLD804-12
Flexible distributor for bottles, BOLA
Closures Bottle Closures
PP screw cap, blue, distributor body and tubing connectors made from PP
  • For bottle thread GLS 80
  • Restricted chemical resistance
  • Temperature resistance from 0 to 110 °C
  • Suitable for vacuum
For drawing or inserting liquids. For use with elastic tubing (e.g. Viton®, Tygon®, silicone). Bent two-step hose connectors on the upper side, straight hose connectors on the bottom side.