Pressure-relief valve with manual ventilation, BOLA
Supplier: Bohlender
E683-18
BOHLE683-18EA 2400 SGD
BOHLE683-18
Pressure-relief valve with manual ventilation, BOLA
Valves Pressure-Relief Valves
Valve made from PPS with flow body made from PTFE. Can be used as a pressure relief valve with adjustable control of pressure or as a valve to prevent a fall in pressure during the filling process. Flow body with 2 GL 18 threaded connectors and valve with toggle for manual venting by tightening, set screw and lock nut to adjust and fix the application pressure in the range from 0,1 to 10 bar (factory setting 1,5 bar).
  • Very good chemical resistance, the fluid medium only comes into contact with PTFE
  • Temperature resistant from −20 to +150°C
  • Pressure
More Product Information