Control valves with GL thread, BOLA
Supplier: Bohlender
E694-18 E694-14
BOHLE694-18EA 2020 SGD
BOHLE694-18 BOHLE694-14
Control valves with GL thread, BOLA
Valves Air Regulation Valves
Two-way valve with straight bore and two connections with GL thread completely made of PTFE.
  • Suitable for vacuum
  • Universal chemical resistance, the flowing product is only exposed to PTFE
Applications include for distributing liquids or gases, manual regulation for constant flow.
Order Now
SPECIFICATIONS
Environmentally Preferable