Screw caps, BOLA
Supplier: Bohlender
H988-40
BOHLH988-40EA 19 SGD
BOHLH988-40
Screw caps, BOLA
Closures Bottle Closures
Screw caps, made of PPS, with handy knurl, with integrated PTFE/silicone gasket for compensation of unevenness on the sealing surface.
  • Universal chemical resistance
  • Temperature resistance: from –20 to +230 °C
After assembly, the product is only exposed to PTFE. The cap has high mechanical and thermal (up to +230 °C) capacitites.