Screw caps high chem, BOLA
Supplier: Bohlender
H998-18
BOHLH998-18EA 127 SGD
BOHLH998-18
Screw caps high chem, BOLA
Closures Bottle Closures
Screw caps made of PP, with handy knurl. Sealing insert made of PTFE with elastic sealing lip and an o-ring for balancing unevenness on the bottle neck.
  • Very good chemical resistance
  • Temperature resistant from 0 to +110 °C
  • Provide tight sealing even at high temperature fluctuations
  • Suitable for vaccum application
Suitable for glass bottles with GLS 80 threads. For the storage of aggressive or pure liquids.