Suction filter, b.safe
Supplier: Bohlender
M515-01
BOHLM515-01EA 225 SGD
BOHLM515-01
Suction filter, b.safe
Filters Liquid Process Filters
Filter candle made of porous PE.
  • With a bore to be put on capillary tubing
  • Restrains pollution from solvents
  • For constant analysis results
Exchange the filter regularly every three months for reliable operation.