Tube tags, b.safe
Supplier: Bohlender
M560-01
BOHLM560-01EA 121 SGD
BOHLM560-01
Tube tags, b.safe
Labels
Flexible tube tag to stick on capillaries and tubes with Ext.Ø 0,4 to 6,35 mm, assorted colours (5 colours per pack).
  • Markable with ball pens
  • Assorted colours
  • Made of high quality PP
Coloured tags to identify capillaries and tubes. Markable with ball pens (removable) and permanent markers, or self-adhesive labels.