Suction filters with PTFE membrane, b.safe
Supplier: Bohlender
M570-05
BOHLM570-05EA 158 SGD
BOHLM570-05
Suction filters with PTFE membrane, b.safe
Filters Liquid Process Filters
Suction Filter with PTFE membrane.
  • With a bore to be put on capillary tubing
  • Restrains pollution from solvents
  • For constant analysis results
Exchange the filter regularly every three months for reliable operation.