Level indicators, b.safe
Supplier: Bohlender
M710-01 M712-01 M715-01 M720-01 M730-01 M732-01 M734-01 M801-01
BOHLM710-01EA 845 SGD
BOHLM710-01 BOHLM712-01 BOHLM715-01 BOHLM720-01 BOHLM730-01 BOHLM732-01 BOHLM734-01 BOHLM801-01
Level indicators, b.safe
Level Sensors
Insert with screw-in thread for barrels or screw cap for canisters. With integrated level indicator to monitor the liquid level during filling as well as a neck with GL 45 thread for the assembly of a b.safe funnel with GL 45 thread.
  • Prevent the waste canisters from overflowing
  • Available for canisters and barrels
  • Suitable for flammable liquid
The red visual pin goes up with increasing filling level. As soon as the visual pin fills the inspection glass completely, the canister is full and has to be exchanged.
Use of a b.safe funnel for filling ofMore Product Information