Waste Tubing Desktop Mounts, b.safe Multiport IV
Supplier: Bohlender
R1127-04
BOHLR1127-04EA 685 SGD
BOHLR1127-04
Waste Tubing Desktop Mounts, b.safe Multiport IV
Tubing Transfer Tubing
One connection for up to four HPLC systems that can be integrated directly in laboratory hoods or working tables. b.safe Waste Tubes lead liquid solvent waste directly into connected canisters.
  • liquid solvent wastes can be led directly from the HPLC device into the collecting canister
  • The single pipe is connected by simply pushing it into the lock nut of the joining piece
  • The combination possibilities for the connectors, like e.g. elbow, pipe bend, allow many different installations
Made of static dissipative PE-EX, suitable for tiles with a thicknessMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable