Antistatic mats, b.safe
Supplier: Bohlender
R630-01
BOHLR630-01EA 675 SGD
BOHLR630-01
Antistatic mats, b.safe
Floor Mats Permanent Floor Mats
Mat made of static dissipative material with permanently mounted grounding cable including clip for earthing connection.
  • Suitable grounding accessories for b.safe products
  • Customised versions can be offered on request
For grounding vessels made of static dissipative materials or persons before handling with easy flammable liquids.
Order Now
SPECIFICATIONS
Environmentally Preferable