Funnels with level control, b.safe
Supplier: Bohlender
R840-15 R840-16 R840-25 R840-26 R880-15 R890-15
BOHLR840-15EA 761 SGD
BOHLR840-15 BOHLR840-16 BOHLR840-25 BOHLR840-26 BOHLR880-15 BOHLR890-15
Funnels with level control, b.safe
Funnels Safety Funnels
b.safe funnels with level control are made of electrically conductive dissipative PE-EX and suitable for use with flammable liquids. Electrically conductive waste canisters are usually black and therefore the filling volume is not visible. This is where the funnel with fill level control comes in.
  • Prevent the waste canisters from overflowing
  • Available for canisters and barrels
  • With lid or splash protector
  • 130 or 180 mm funnel diameter
The funnels are available in two sizes, each with a hinged lid or splash guard. The unique design with slanted threadMore Product Information