Desiccator cabinets, SICCO Mini Basic series
Supplier: Bohlender
V1850-01 V1850-02 V1850-03
BOHLV1850-01EA 479 SGD
BOHLV1850-01 BOHLV1850-02 BOHLV1850-03
Desiccator cabinets, SICCO Mini Basic series
Desiccators Desiccator Cabinets
Clear, polycarbonate. Each compartment has one door with snap fit closing and integrated gasket ensuring easy opening and tight closure. Temperature resistant from −35 to +70 °C, suitable for use under normal pressure.
  • Large opening and door with wide opening angle enable user to access the entire interior space
  • Robust construction, lightweight, portable and stackable
  • Supplied with desiccant which is in a handy, resealable container
Ordering information: Shelves must be ordered separately.