Desiccator cabinets, SICCO Maxi series
Supplier: Bohlender
V1852-01 V1853-01
BOHLV1852-01EA 9880 SGD
BOHLV1852-01 BOHLV1853-01
Desiccator cabinets, SICCO Maxi series
Desiccators Desiccator Cabinets
Transparent acrylic. Cabinets have a reinforced aluminium frame, door with foam rubber gasket and magnetic strip. Designed for storing or drying humidity sensitive products using silica gel. The controlled environment inside the desiccator is ideal for storing reference materials, retained samples, metallographic specimens, tobacco and DNA samples.
  • Door mounted electronic hygrometer* indicates minimum and maximum temperature and humidity values
  • Temperature resistance from −20 °C to +70 °C
  • Easy to move about as cabinets have four castors, two of them with
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable