Desiccator cabinets, SICCO maxi series
Supplier: Bohlender
V1927-07
BOHLV1927-07EA 13120 SGD
BOHLV1927-07
Desiccator cabinets, SICCO maxi series
Desiccators Desiccator Cabinets
Orange acrylic cabinets have a reinforced aluminium frame, door with foam rubber gasket and magnetic strip, orange acrylic panels reduce light incidence and protect against ultraviolet radiation. Especially suitable for storage of light-sensitive substances
  • Door mounted electronic hygrometer indicates minimum and maximum temperature and humidity values
  • Temperature resistance from −20 to +70 °C
  • Easy to move about as cabinets have four castors
Load capacity: 30 kg/shelf, 160 kg (total)
Load capacity: 30 kg/shelf, 160 kg (total)