Vacuum trap, serrated connections
Supplier: CHEMGLASS
CG-4515-20
CGLACG-4515-20EA 888 SGD
CGLACG-4515-20
Vacuum trap, serrated connections
Vacuum Traps
Two-piece construction for easy removal of condensate. Inlet and outlet are 180° apart for installation into a variety of vacuum systems.
  • Stopper has a standard taper inner joint and body has an outer joint
  • Body length is the overall length from the top of the joint to the bottom of the round bottom
  • Serrated hose connections have an O.D. of 13 mm at the smallest serration and 16 mm at the largest serration