Guaiacol detection kit
Supplier: DOEHLER GROUP
204737991
DOEH204737991EA 512 SGD
DOEH204737991
Guaiacol detection kit
Media for Microbiology Prepared Media for Microbiology
Enzymatic test kit for the detection of off-flavour forming Alicyclobacillus. Guaiacol Detection Kit is a reliable and sensitive enzymatic detection of Guaiacol-forming Alicyclobacilli. When Guaiacol-forming bacteria are present, the transparent to slightly yellow Guaiacol is transformed into the clearly-visible, brownish complex Tetraguaiacol with the enzyme peroxidase from the Detection Kit.
  • Comprehensive Detection Kit including BAT Broth in tubes and Guaiacol Standard Solution
  • Simple visual analysis without additional technical equipment