HLP Medium (Hsu's Lactobacillus/Pediococcus Medium)
Supplier: DOEHLER GROUP
923556244
DOEH923556244EA 445 SGD
DOEH923556244
HLP Medium (Hsu's Lactobacillus/Pediococcus Medium)
Media for Microbiology Prepared Media for Microbiology
Siebel-Instituts nutrient medium for the selective detection and quantitative determination of Lactobacillus and Pediococcus from the brewing environment.
  • Certified by the Siebel-Institute for technology
  • Ready-to-use powder to prepare HLP gel or agar without prior autoclaving
  • Functionally tested with strains originally isolated from the brewing environment
HLP promote the growth of beer spoiling bacteria like Lactobacillus spp./Pedicococcus spp. which are commonly encountered in a brewery. Cycloheximide contained in the medium inhibits the growth of culture yeast.
Order Now
SPECIFICATIONS
Environmentally Preferable