Protein quantitation assay kits, Non-Interfering™ (NI™) Protein Assay
Supplier: G-Biosciences
786-005
GENO786-005EA 1260 SGD
GENO786-005
Protein quantitation assay kits, Non-Interfering™ (NI™) Protein Assay
Assays Protein Assays
A highly sensitive colorimetric protein assay that overcomes interference of common laboratory agents present in protein solutions and shows minimal protein-to-protein variation. The assay is unaffected by the presence of common laboratory agents such as reducing agents, chelating agents, detergents, amines, sugars, chaotropes, salts, drugs, antibiotics, cobalt, and others.
Order Now
SPECIFICATIONS
Environmentally Preferable