Syringes for HPLC autosamplers, for Spectra-Physics
Supplier: HAMILTON BONADUZ
202192
HAMI202192EA 334 SGD
HAMI202192
Syringes for HPLC autosamplers, for Spectra-Physics
Syringes Syringes for Chromatography
These gas tight syringes are designed for HPLC autosamplers.
  • Temperature range: 20 to 50 °C
  • Thread size 1/4-28