Syringes, Neuros™
Supplier: HAMILTON BONADUZ
65460-02 65457-02 65459-02 65460-03
HAMI65460-02EA 1490 SGD
HAMI65460-02 HAMI65457-02 HAMI65459-02 HAMI65460-03
Syringes, Neuros™
Syringes Syringes for Chromatography
Developed specifically for the neurosciences, these syringes minimise injection site damage by increasing needle rigidity and delivering a micro volume of liquid to an exact location.
  • Compatibility with most infusion pumps and stereotaxic holders
  • Black needle sleeve indicates the exposed needle length
The technology is offered on several models from the 700 Microliter™, 1700 Gastight® and 7000 Modified Microliter™ series. These syringes have very little to no dead volume which preserves the maximum amount of buffer and solution. They feature twoMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable