600 series Microliter™ syringes
Supplier: HAMILTON BONADUZ
7632-01 7633-01
HAMI7632-01EA 531 SGD
HAMI7632-01 HAMI7633-01
600 series Microliter™ syringes
Syringes Syringes for Chromatography
The 600 series syringes are a half stroke version of the original 700 series syringes. This series of syringes is ideal for dispensing volumes from 0,25 μl up to 5 μl. The 600 series is often used in applications that require one handed operation because of the short stroke length and the reinforced plunger.