900 series Microliter™ syringes
Supplier: HAMILTON BONADUZ
87925 7648-01 80360
HAMI87925EA 525 SGD
HAMI87925 HAMI7648-01 HAMI80360
900 series Microliter™ syringes
Syringes Syringes for Chromatography
The 900 series syringe is the economical version of the 800 series syringe. This series of syringes is ideal for dispensing volumes from 0,5 μl up to 10 μl. The syringe holder is permanently attached to the syringe barrel. This reduces the initial cost of the syringe but eliminates the possibility of replacement components.
Syringe terminations:
N, Cemented Needle
For low volume syringes the needles are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. NotMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable